siRNA Details
| Gene Symbol: CCDC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135197.1 | NP_001128669.1 | 6 | 568 | ||
| NM_178173.3 | NP_835467.2 | 8 | 866 | |||
| XM_006713124.3 | XP_006713187.1 | 6 | 743 | |||
| XM_011533670.2 | XP_011531972.1 | 5 | 577 | |||
| XM_011533671.2 | XP_011531973.1 | 5 | 571 | |||
| XM_011533672.2 | XP_011531974.1 | 6 | 936 | |||
| XM_011533673.2 | XP_011531975.1 | 6 | 968 | |||
| GenBank mRNA | AK058049.1 | - | 8 | 757 | ||
| AK303033.1 | - | 6 | 568 | |||
| BC036202.1 | - | 8 | 866 | 4836976 | ||

CCDC36
Genomic Map