siRNA Details
| Gene Symbol: ST8SIA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304450.1 | NP_001291379.1 | 4 | 1481 | ||
| NM_003034.3 | NP_003025.1 | 5 | 1591 | |||
| GenBank mRNA | AK290941.1 | - | 5 | 1232 | ||
| AK297310.1 | - | 4 | 964 | |||
| BC117285.1 | - | 5 | 1510 | 40125836 | ||
| BC126162.1 | - | 5 | 1510 | 8991878 | ||
| D26360.1 | - | 5 | 1212 | |||
| EU041716.1 | - | 4 | 1481 | |||
| L32867.1 | - | 5 | 1176 | |||
| L43494.1 | - | 5 | 1120 | |||
| X77922.1 | - | 5 | 1591 | |||

ST8SIA1
Genomic Map