siRNA Details
| Gene Symbol: DAW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178821.1 | NP_849143.1 | 2, 3 | 200 | ||
| XM_011510755.1 | XP_011509057.1 | 3, 4 | 809 | |||
| GenBank mRNA | AK097409.1 | - | 2, 3 | 200 | ||
| AK098821.1 | - | 2, 3 | 148 | |||
| AK127882.1 | - | 2, 3 | 757 | |||
| AK301861.1 | - | 3, 4 | 200 | |||
| BC036377.2 | - | 2, 3 | 180 | 4824989 | ||

DAW1
Genomic Map