siRNA Details
| Gene Symbol: ZNF233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207005.1 | NP_001193934.1 | 5 | 2426 | ||
| NM_181756.2 | NP_861421.2 | 5 | 2456 | |||
| XM_017026758.1 | XP_016882247.1 | 5 | 2828 | |||
| XM_017026759.1 | XP_016882248.1 | 5 | 2609 | |||
| GenBank mRNA | AK055310.1 | - | 5 | 2608 | ||
| AK095351.1 | - | 5 | 2447 | |||
| CR743367.1 | - | 1 | 244 | |||

ZNF233
Genomic Map