siRNA Details
| Gene Symbol: C2orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006711859.3 | XP_006711922.1 | 4 | 1161 | ||
| GenBank mRNA | AK057872.1 | - | 3 | 2105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf50| Gene Symbol: C2orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006711859.3 | XP_006711922.1 | 4 | 1161 | ||
| GenBank mRNA | AK057872.1 | - | 3 | 2105 | ||