siRNA Details
| Gene Symbol: IZUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321865.1 | NP_001308794.1 | 1 | 276 | ||
| NM_182575.2 | NP_872381.2 | 2 | 751 | |||
| XM_005258793.4 | XP_005258850.1 | 2 | 484 | |||
| XM_011526811.2 | XP_011525113.1 | 1 | 1255 | |||
| XM_011526812.2 | XP_011525114.1 | 2 | 831 | |||
| XM_011526813.2 | XP_011525115.1 | 2 | 680 | |||
| XM_011526815.2 | XP_011525117.1 | 1 | 1255 | |||
| XM_017026646.1 | XP_016882135.1 | 1 | 1255 | |||
| GenBank mRNA | AB195682.1 | - | 2 | 609 | ||
| AK301750.1 | - | 1 | 1255 | |||
| AY552609.1 | - | 1 | 203 | |||
| BC034769.1 | - | 2 | 751 | 5167853 | ||

IZUMO1
Genomic Map