siRNA Details
| Gene Symbol: IZUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321864.1 | NP_001308793.1 | 9 | 906 | ||
| NM_001321865.1 | NP_001308794.1 | 9 | 1069 | |||
| NM_182575.2 | NP_872381.2 | 10 | 1507 | |||
| NR_135832.1 | 9 | 995 | ||||
| XM_005258793.4 | XP_005258850.1 | 10 | 1240 | |||
| XM_011526811.2 | XP_011525113.1 | 8 | 1930 | |||
| XM_011526812.2 | XP_011525114.1 | 10 | 1587 | |||
| XM_011526813.2 | XP_011525115.1 | 10 | 1436 | |||
| XM_011526815.2 | XP_011525117.1 | 9 | 2150 | |||
| XM_017026646.1 | XP_016882135.1 | 9 | 2011 | |||
| GenBank mRNA | AB195682.1 | - | 10 | 1365 | ||
| AK301750.1 | - | 9 | 2150 | |||
| AY552609.1 | - | 9 | 996 | |||
| AY552610.1 | - | 9 | 922 | |||
| BC034769.1 | - | 10 | 1507 | 5167853 | ||

IZUMO1
Genomic Map