siRNA Details
| Gene Symbol: LINC00663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026956.1 | 3 | 505 | |||
| GenBank mRNA | AK022418.1 | - | 1 | 139 | ||
| AK289657.1 | - | 3 | 485 | |||
| BC034236.1 | - | 4 | 637 | |||
| DB196080.1 | - | 3 | 505 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00663| Gene Symbol: LINC00663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026956.1 | 3 | 505 | |||
| GenBank mRNA | AK022418.1 | - | 1 | 139 | ||
| AK289657.1 | - | 3 | 485 | |||
| BC034236.1 | - | 4 | 637 | |||
| DB196080.1 | - | 3 | 505 | |||