siRNA Details
| Gene Symbol: C20orf203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182584.2 | NP_872390.2 | 3 | 1812 | ||
| GenBank mRNA | AK091025.1 | - | 3 | 1709 | ||
| GU931819.1 | - | 4 | 2122 | |||
| GU931820.1 | - | 4 | 2122 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf203| Gene Symbol: C20orf203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182584.2 | NP_872390.2 | 3 | 1812 | ||
| GenBank mRNA | AK091025.1 | - | 3 | 1709 | ||
| GU931819.1 | - | 4 | 2122 | |||
| GU931820.1 | - | 4 | 2122 | |||