siRNA Details
| Gene Symbol: SAMD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304366.1 | NP_001291295.1 | 6 | 1217 | ||
| NM_182610.3 | NP_872416.1 | 6 | 1240 | |||
| NR_130713.1 | 7 | 1330 | ||||
| GenBank mRNA | BC117339.1 | - | 5 | 924 | 40125890 | |
| BC143909.1 | - | 6 | 1036 | |||
| BX537903.1 | - | 6 | 1120 | |||
| BX648534.1 | - | 6 | 1098 | |||
| BX648989.1 | - | 6 | 1120 | |||
| BX648991.1 | - | 6 | 1218 | |||

SAMD7
Genomic Map