siRNA Details
| Gene Symbol: ANKRD18DP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003291.1 | 10 | 1453 | |||
| GenBank mRNA | AK125940.1 | - | 1 | 3506 | ||
| BC053879.1 | - | 6 | 1249 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD18DP| Gene Symbol: ANKRD18DP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003291.1 | 10 | 1453 | |||
| GenBank mRNA | AK125940.1 | - | 1 | 3506 | ||
| BC053879.1 | - | 6 | 1249 | |||