siRNA Details
| Gene Symbol: RPS19BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194326.3 | NP_919307.1 | 4 | 846 | ||
| NR_130151.1 | 5 | 937 | ||||
| GenBank mRNA | AK025423.1 | - | 4 | 739 | ||
| AK055865.1 | - | 2 | 2724 | |||
| BC037573.1 | - | 4 | 856 | 5399759 | ||
| BC047711.1 | - | 4 | 738 | 6064917 | ||
| BQ675376.1 | - | 2 | 478 | |||
| CR456443.1 | - | 4 | 776 | |||

RPS19BP1
Genomic Map