siRNA Details
| Gene Symbol: RINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195833.1 | NP_001182762.1 | 10 | 1514 | ||
| NM_198445.3 | NP_940847.1 | 10 | 1498 | |||
| XM_006723015.2 | XP_006723078.1 | 10 | 1508 | |||
| XM_006723016.2 | XP_006723079.1 | 10 | 1476 | |||
| XM_011526454.2 | XP_011524756.1 | 10 | 1578 | |||
| XM_011526455.2 | XP_011524757.1 | 11 | 1412 | |||
| GenBank mRNA | AK124506.1 | - | 2 | 122 | ||
| AK126119.1 | - | 7 | 1894 | |||
| AK127808.1 | - | 10 | 1473 | |||
| AK298327.1 | - | 10 | 1514 | |||
| BU196609.1 | - | 1 | 68 | |||

RINL
Genomic Map