siRNA Details
| Gene Symbol: CXCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198477.2 | NP_940879.1 | 4 | 644 | ||
| NR_133910.1 | 4 | 661 | ||||
| GenBank mRNA | AK292985.1 | - | 4 | 542 | ||
| AY358433.1 | - | 4 | 580 | |||
| AY598464.1 | - | 4 | 644 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXCL17| Gene Symbol: CXCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198477.2 | NP_940879.1 | 4 | 644 | ||
| NR_133910.1 | 4 | 661 | ||||
| GenBank mRNA | AK292985.1 | - | 4 | 542 | ||
| AY358433.1 | - | 4 | 580 | |||
| AY598464.1 | - | 4 | 644 | |||