siRNA Details
| Gene Symbol: TMEM110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198563.2 | NP_940965.1 | 8 | 1091 | ||
| GenBank mRNA | AY726591.1 | - | 8 | 854 | ||
| BC022209.1 | - | 6 | 718 | |||
| BC047015.1 | - | 8 | 1041 | 5199769 | ||
| BX648562.1 | - | 2 | 298 | |||

TMEM110
Genomic Map