siRNA Details
| Gene Symbol: RBM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080504.2 | NP_001073973.2 | 11, 12 | 2672 | ||
| XM_005246075.1 | XP_005246132.1 | 11, 12 | 3013 | |||
| XM_011511155.1 | XP_011509457.1 | 3, 4 | 181 | |||
| XM_017004054.1 | XP_016859543.1 | 11, 12 | 3016 | |||
| XM_017004055.1 | XP_016859544.1 | 11, 12 | 3386 | |||
| XM_017004056.1 | XP_016859545.1 | 11, 12 | 2956 | |||
| XM_017004058.1 | XP_016859547.1 | 6, 7 | 1354 | |||
| GenBank mRNA | AK130316.1 | - | 2, 3 | 111 | ||
| BC126111.1 | - | 8, 9 | 755 | |||

RBM44
Genomic Map