siRNA Details
| Gene Symbol: RBM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080504.2 | NP_001073973.2 | 14 | 3084 | ||
| XM_005246075.1 | XP_005246132.1 | 14 | 3425 | |||
| XM_011511155.1 | XP_011509457.1 | 6 | 593 | |||
| XM_017004054.1 | XP_016859543.1 | 14 | 3428 | |||
| XM_017004055.1 | XP_016859544.1 | 14 | 3798 | |||
| XM_017004056.1 | XP_016859545.1 | 14 | 3368 | |||
| XM_017004058.1 | XP_016859547.1 | 9 | 1766 | |||
| GenBank mRNA | AK130316.1 | - | 5 | 523 | ||
| BC126111.1 | - | 11 | 1167 | |||
| BX648107.1 | - | 13 | 3737 | |||

RBM44
Genomic Map