siRNA Details
| Gene Symbol: RBM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080504.2 | NP_001073973.2 | 14 | 3116 | ||
| XM_005246075.1 | XP_005246132.1 | 14 | 3457 | |||
| XM_011511155.1 | XP_011509457.1 | 6 | 625 | |||
| XM_017004054.1 | XP_016859543.1 | 14 | 3460 | |||
| XM_017004055.1 | XP_016859544.1 | 14 | 3830 | |||
| XM_017004056.1 | XP_016859545.1 | 14 | 3400 | |||
| XM_017004058.1 | XP_016859547.1 | 9 | 1798 | |||
| GenBank mRNA | AK130316.1 | - | 5 | 555 | ||
| BC126111.1 | - | 11 | 1199 | |||
| BX648107.1 | - | 13 | 3769 | |||

RBM44
Genomic Map