siRNA Details
| Gene Symbol: FAM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004699.3 | NP_004690.1 | 5 | 612 | ||
| GenBank mRNA | AD001530.1 | - | 5 | 577 | ||
| AK313697.1 | - | 5 | 560 | |||
| BC000028.2 | - | 5 | 583 | 3505365 | ||
| CR992165.1 | - | 5 | 612 | |||
| D83260.1 | - | 5 | 482 | |||

FAM50A
Genomic Map