siRNA Details
| Gene Symbol: SLU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006425.4 | NP_006416.3 | 5 | 599 | ||
| XM_005265793.3 | XP_005265850.1 | 5 | 946 | |||
| XM_011534418.2 | XP_011532720.1 | 6 | 649 | |||
| GenBank mRNA | AF101074.1 | - | 4 | 444 | ||
| AK024875.1 | - | 5 | 519 | |||
| AK314564.1 | - | 5 | 519 | |||
| BC010634.1 | - | 5 | 525 | 3871882 | ||
| CA976106.1 | - | 3 | 190 | |||
| DA409854.1 | - | 5 | 525 | |||

SLU7
Genomic Map