siRNA Details
| Gene Symbol: SLU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006425.4 | NP_006416.3 | 8 | 960 | ||
| XM_005265793.3 | XP_005265850.1 | 8 | 1307 | |||
| XM_011534418.2 | XP_011532720.1 | 9 | 1010 | |||
| GenBank mRNA | AF101074.1 | - | 7 | 805 | ||
| AK024875.1 | - | 8 | 880 | |||
| AK314564.1 | - | 8 | 880 | |||
| BC010634.1 | - | 8 | 886 | 3871882 | ||
| CA976106.1 | - | 6 | 551 | |||

SLU7
Genomic Map