siRNA Details
| Gene Symbol: WDR45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029896.1 | NP_001025067.1 | 11 | 1345 | ||
| NM_007075.3 | NP_009006.2 | 12 | 1615 | |||
| GenBank mRNA | AJ005897.1 | - | 11 | 1301 | ||
| AK023344.1 | - | 9 | 2157 | |||
| AK225122.1 | - | 11 | 1324 | |||
| BC000464.2 | - | 11 | 1339 | 2822742 | ||
| BC003037.1 | - | 11 | 1339 | 2822742 | ||
| BC009027.1 | - | 8 | 1002 | |||
| BC069206.1 | - | 12 | 1615 | 5801768 | ||

WDR45
Genomic Map