siRNA Details
| Gene Symbol: FAM50B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012135.2 | NP_036267.1 | 2 | 952 | ||
| XM_017010729.1 | XP_016866218.1 | 2 | 935 | |||
| GenBank mRNA | BC001261.1 | - | 2 | 930 | 3457886 | |
| Y18503.1 | - | 2 | 920 | |||

FAM50B
Genomic Map