siRNA Details
Gene Symbol: DROSHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100412.1 | NP_001093882.1 | 12 | 2139 | ||
NM_013235.4 | NP_037367.3 | 12 | 2174 | |||
XM_005248291.3 | XP_005248348.1 | 13 | 2250 | |||
XM_005248292.3 | XP_005248349.1 | 13 | 2250 | |||
XM_005248293.3 | XP_005248350.1 | 12 | 2157 | |||
XM_005248294.3 | XP_005248351.1 | 11 | 2046 | |||
XM_011514033.2 | XP_011512335.1 | 13 | 2250 | |||
XM_017009399.1 | XP_016864888.1 | 12 | 2157 | |||
XM_017009400.1 | XP_016864889.1 | 12 | 2139 | |||
XM_017009401.1 | XP_016864890.1 | 11 | 2046 | |||
GenBank mRNA | AB209501.1 | - | 5 | 9769 | ||
AF189011.1 | - | 12 | 2060 | |||
AJ242976.1 | - | 7 | 1118 | |||
AK308093.1 | - | 11 | 1948 | |||
BC054003.1 | - | 11 | 1951 | 6148714 | ||
BX647724.1 | - | 12 | 2042 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001130149, NM_026799, AF533013, BC055696, AK077549, and others.
Panther Classification:
Gene Ontology Categories:
