siRNA Details
| Gene Symbol: FAM131B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031690.2 | NP_001026860.2 | 7 | 1218 | ||
| NM_001278297.1 | NP_001265226.1 | 6 | 1124 | |||
| NM_014690.4 | NP_055505.3 | 7 | 1111 | |||
| XM_005250073.3 | XP_005250130.1 | 7 | 1432 | |||
| XM_006716186.2 | XP_006716249.1 | 7 | 1210 | |||
| GenBank mRNA | AB018316.1 | - | 6 | 1006 | ||
| AK124557.1 | - | 7 | 1162 | |||
| AK291470.1 | - | 7 | 1083 | |||
| AL832579.2 | - | 6 | 2699 | |||
| BC045611.1 | - | 7 | 1223 | 5300056 | ||
| BC050543.1 | - | 7 | 1086 | 5768309 | ||

FAM131B
Genomic Map