siRNA Details
| Gene Symbol: NSA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014886.4 | NP_055701.1 | 5 | 950 | ||
| NR_073403.1 | 6 | 1059 | ||||
| XM_011543098.1 | XP_011541400.1 | 5 | 628 | |||
| GenBank mRNA | AF077615.1 | - | 5 | 664 | ||
| AF372458.1 | - | 5 | 675 | |||
| AJ012409.1 | - | 5 | 662 | |||
| AK313784.1 | - | 5 | 693 | |||
| AY443419.1 | - | 4 | 580 | |||
| BC005288.1 | - | 5 | 693 | 3683720 | ||

NSA2
Genomic Map