siRNA Details
| Gene Symbol: ETNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039481.1 | NP_001034570.1 | 2 | 573 | ||
| NM_018638.4 | NP_061108.2 | 2 | 573 | |||
| XM_005253420.4 | XP_005253477.1 | 2 | 570 | |||
| XM_017019580.1 | XP_016875069.1 | 2 | 570 | |||
| XM_017019581.1 | XP_016875070.1 | 2 | 570 | |||
| GenBank mRNA | AF207600.2 | - | 2 | 639 | ||
| AK074425.1 | - | 2 | 518 | |||
| AL137750.1 | - | 2 | 523 | |||
| BC006111.1 | - | 2 | 525 | |||
| BC066907.1 | - | 2 | 573 | 5260109 | ||
| BT007084.1 | - | 2 | 284 | |||

ETNK1
Genomic Map