siRNA Details
| Gene Symbol: TECPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015395.2 | NP_056210.1 | 11 | 1605 | ||
| XM_005250253.3 | XP_005250310.1 | 11 | 1577 | |||
| XM_017011937.1 | XP_016867426.1 | 10 | 1487 | |||
| XM_017011938.1 | XP_016867427.1 | 11 | 1589 | |||
| GenBank mRNA | AB037779.1 | - | 9 | 1181 | ||
| AK057048.1 | - | 10 | 1332 | |||
| AK292996.1 | - | 11 | 1507 | |||
| AL117495.1 | - | 1 | 55 | |||
| BC053591.1 | - | 11 | 1510 | 5502429 | ||

TECPR1
Genomic Map