siRNA Details
| Gene Symbol: KIAA1429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015496.4 | NP_056311.2 | 7 | 773 | ||
| NM_183009.2 | NP_892121.1 | 7 | 773 | |||
| XM_017013306.1 | XP_016868795.1 | 6 | 553 | |||
| XM_017013307.1 | XP_016868796.1 | 6 | 596 | |||
| XM_017013308.1 | XP_016868797.1 | 5 | 586 | |||
| GenBank mRNA | AB037850.1 | - | 7 | 697 | ||
| AF116724.1 | - | 7 | 697 | |||
| AL832487.1 | - | 7 | 715 | |||
| BC112288.1 | - | 7 | 785 | 8327756 | ||
| BC113380.1 | - | 7 | 773 | 8322432 | ||

KIAA1429
Genomic Map