siRNA Details
| Gene Symbol: SNRPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003090.3 | NP_003081.2 | 6, 7 | 639 | ||
| GenBank mRNA | AK223070.1 | - | 6, 7 | 596 | ||
| AK223110.1 | - | 6, 7 | 561 | |||
| AK312200.1 | - | 6, 7 | 570 | |||
| BC022816.1 | - | 6, 7 | 592 | 4904205 | ||
| BC071717.1 | - | 1 | 547 | 4514975 | ||
| X13482.1 | - | 6, 7 | 591 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021336, BC013777, AK206837, BC029642, AK153502, and others.

SNRPA1
Genomic Map