siRNA Details
Gene Symbol: NOC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015658.3 | NP_056473.2 | 16 | 1914 | ||
GenBank mRNA | AK022756.1 | - | 10 | 1157 | ||
AK024284.1 | - | 16 | 1868 | |||
AK092491.1 | - | 2 | 711 | |||
AK225239.1 | - | 16 | 1869 | |||
AK315080.1 | - | 16 | 1883 | |||
AL050019.1 | - | 16 | 1870 | |||
BC003555.1 | - | 16 | 1887 | 3546209 |
Gene Symbol: NOC2LP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002826.2 | 1 | 1859 | |||
GenBank mRNA | AK302229.1 | - | 1 | 1859 | ||
BC060851.1 | - | 1 | 1852 |
