siRNA Details
| Gene Symbol: GIMAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015660.2 | NP_056475.1 | 3 | 886 | ||
| GenBank mRNA | AK074199.1 | - | 3 | 885 | ||
| AL110151.1 | - | 3 | 865 | |||
| BC013934.1 | - | 3 | 886 | 3950542 | ||
| BC032345.1 | - | 3 | 862 | 5208411 | ||

GIMAP2
Genomic Map