siRNA Details
| Gene Symbol: WWC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015691.3 | NP_056506.2 | 3, 4 | 424 | ||
| GenBank mRNA | AB033106.1 | - | 1, 2 | 47 | ||
| AK292961.1 | - | 2, 3 | 306 | |||
| BU500300.1 | - | 3, 4 | 632 | |||
| BU500742.1 | - | 3, 4 | 608 | |||
| KC987947.1 | - | 3, 4 | 601 | |||

WWC3
Genomic Map