siRNA Details
| Gene Symbol: ZNF706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042510.1 | NP_001035975.1 | 5 | 1453 | ||
| NM_001267708.1 | NP_001254637.1 | 6 | 1497 | |||
| NM_001267709.1 | NP_001254638.1 | 4 | 1251 | |||
| NM_016096.3 | NP_057180.1 | 4 | 1338 | |||
| GenBank mRNA | AK129859.1 | - | 1 | 202 | ||
| BC068524.1 | - | 4 | 1221 | 5263977 | ||

ZNF706
Genomic Map