siRNA Details
| Gene Symbol: HERC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016323.3 | NP_057407.2 | 13 | 1866 | ||
| XM_011532022.2 | XP_011530324.2 | 13 | 2379 | |||
| GenBank mRNA | AB027289.1 | - | 13 | 1826 | ||
| AK093155.1 | - | 3 | 1201 | |||
| AK301756.1 | - | 3 | 434 | |||
| AK302142.1 | - | 7 | 738 | |||
| AY337518.1 | - | 13 | 1866 | |||
| BC140716.1 | - | 13 | 1759 | 9021584 | ||

HERC5
Genomic Map