siRNA Details
Gene Symbol: SOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003102.2 | NP_003093.2 | 2 | 1163 | ||
GenBank mRNA | BC014418.1 | - | 2 | 1028 | 4644224 | |
J02947.1 | - | 2 | 1027 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003102.2 | NP_003093.2 | 2 | 1163 | ||
GenBank mRNA | BC014418.1 | - | 2 | 1028 | 4644224 | |
J02947.1 | - | 2 | 1027 |