siRNA Details
| Gene Symbol: HGH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016458.3 | NP_057542.2 | 5 | 1061 | ||
| GenBank mRNA | AF151022.1 | - | 2 | 453 | ||
| AK098226.1 | - | 6 | 755 | |||
| BC003035.1 | - | 5 | 1061 | 2822563 | ||
| BC009915.2 | - | 5 | 1061 | 2822563 | ||
| BC015471.1 | - | 6 | 592 | |||

HGH1
Genomic Map