siRNA Details
| Gene Symbol: PART1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028508.1 | 2 | 1665 | |||
| GenBank mRNA | AF163475.1 | - | 2 | 1665 | ||
| BQ898163.1 | - | 1 | 483 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PART1| Gene Symbol: PART1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028508.1 | 2 | 1665 | |||
| GenBank mRNA | AF163475.1 | - | 2 | 1665 | ||
| BQ898163.1 | - | 1 | 483 | |||