siRNA Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300783.1 | NP_001287712.1 | 2 | 827 | ||
| NM_001308087.1 | NP_001295016.1 | 2 | 540 | |||
| NM_001308088.1 | NP_001295017.1 | 4 | 780 | |||
| NM_016644.2 | NP_057728.1 | 3 | 952 | |||
| XM_011543452.2 | XP_011541754.1 | 3 | 2439 | |||
| XM_011543453.2 | XP_011541755.1 | 3 | 921 | |||
| XM_011543454.2 | XP_011541756.1 | 4 | 1027 | |||
| GenBank mRNA | AF242769.1 | - | 3 | 906 | ||
| BC038838.1 | - | 3 | 639 | 5170587 | ||
| BC092474.1 | - | 2 | 828 | 30528467 | ||
| DA328660.1 | - | 2 | 541 | |||

PRR16
Genomic Map