siRNA Details
| Gene Symbol: PLXNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017514.4 | NP_059984.3 | 22 | 4035 | ||
| XM_005274705.4 | XP_005274762.2 | 22 | 3975 | |||
| XM_005274706.4 | XP_005274763.2 | 22 | 3707 | |||
| XM_006724829.3 | XP_006724892.1 | 22 | 3939 | |||
| XM_011531183.2 | XP_011529485.1 | 21 | 3399 | |||
| GenBank mRNA | BG820285.1 | - | 4 | 467 | ||
| X87852.1 | - | 22 | 4044 | |||

PLXNA3
Genomic Map