siRNA Details
| Gene Symbol: SAMD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193307.1 | NP_001180236.1 | 2 | 2510 | ||
| NM_017654.3 | NP_060124.2 | 3 | 2611 | |||
| GenBank mRNA | AB095925.2 | - | 3 | 2545 | ||
| AF445355.1 | - | 3 | 2611 | |||
| AK122951.1 | - | 2 | 2448 | |||
| AK125131.1 | - | 1 | 660 | |||
| BC132773.1 | - | 2 | 2441 | 40146795 | ||
| BC132775.1 | - | 2 | 2441 | 40146797 | ||
| BC150249.1 | - | 3 | 2566 | 9007474 | ||
| BX647072.1 | - | 3 | 2567 | |||

SAMD9
Genomic Map