siRNA Details
| Gene Symbol: ZNF280C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017666.4 | NP_060136.1 | 13 | 1613 | ||
| XM_006724765.3 | XP_006724828.1 | 12 | 1417 | |||
| GenBank mRNA | AK000102.1 | - | 13 | 1536 | ||
| AK290373.1 | - | 13 | 1564 | |||
| AL133204.1 | - | 12 | 1401 | |||
| AL834333.1 | - | 13 | 1544 | |||
| BC051728.1 | - | 13 | 1564 | 4828435 | ||

ZNF280C
Genomic Map