siRNA Details
| Gene Symbol: GIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017676.2 | NP_060146.2 | 3 | 340 | ||
| NR_134299.1 | 3 | 340 | ||||
| XM_005272028.3 | XP_005272085.1 | 3 | 351 | |||
| XM_017009599.1 | XP_016865088.1 | 3 | 1775 | |||
| GenBank mRNA | AK000132.1 | - | 3 | 338 | ||
| AK225572.1 | - | 3 | 337 | |||
| AK295796.1 | - | 3 | 328 | |||
| BC015325.1 | - | 3 | 299 | |||
| BC151135.1 | - | 2 | 280 | 9057035 | ||
| BC157835.1 | - | 2 | 280 | 9057054 | ||
| DB101499.1 | - | 3 | 340 | |||

GIN1
Genomic Map