siRNA Details
| Gene Symbol: GIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317954.1 | NP_001304883.1 | 7 | 1335 | ||
| NM_017676.2 | NP_060146.2 | 8 | 1529 | |||
| NR_134299.1 | 5 | 874 | ||||
| XM_005272028.3 | XP_005272085.1 | 8 | 1536 | |||
| XM_017009599.1 | XP_016865088.1 | 8 | 2964 | |||
| GenBank mRNA | AK000132.1 | - | 8 | 1526 | ||
| AK225572.1 | - | 8 | 1526 | |||
| AK295796.1 | - | 8 | 1517 | |||
| BC015325.1 | - | 5 | 833 | |||
| BC151135.1 | - | 7 | 1469 | 9057035 | ||
| BC157835.1 | - | 7 | 1469 | 9057054 | ||
| CR627424.1 | - | 7 | 1306 | |||

GIN1
Genomic Map