siRNA Details
| Gene Symbol: INTS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017864.3 | NP_060334.2 | 17 | 2384 | ||
| NR_073444.1 | 17 | 2384 | ||||
| NR_073445.1 | 17 | 2353 | ||||
| XM_017013616.1 | XP_016869105.1 | 17 | 2384 | |||
| XM_017013618.1 | XP_016869107.1 | 14 | 1937 | |||
| XM_017013619.1 | XP_016869108.1 | 17 | 2343 | |||
| GenBank mRNA | AB161944.1 | - | 17 | 2368 | ||
| AK000537.1 | - | 7 | 938 | |||
| AK001403.1 | - | 10 | 1324 | |||
| AK091278.1 | - | 16 | 2121 | |||
| AK225533.1 | - | 7 | 938 | |||
| BC050536.1 | - | 9 | 1172 | |||
| BC064915.1 | - | 9 | 1213 | |||
| BC136754.1 | - | 17 | 2298 | 9020744 | ||
| BC144396.1 | - | 17 | 2298 | |||
| BC144397.1 | - | 17 | 2267 | |||
| BX538203.1 | - | 13 | 1707 | |||

INTS8
Genomic Map