siRNA Details
| Gene Symbol: WDR70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018034.2 | NP_060504.1 | 13 | 1437 | ||
| XM_005248318.3 | XP_005248375.1 | 13 | 1455 | |||
| XM_011514061.2 | XP_011512363.1 | 7 | 750 | |||
| XM_017009604.1 | XP_016865093.1 | 12 | 1271 | |||
| XM_017009605.1 | XP_016865094.1 | 12 | 1238 | |||
| GenBank mRNA | AK001095.1 | - | 13 | 1379 | ||
| AK291179.1 | - | 13 | 1459 | |||
| AL512685.1 | - | 13 | 1438 | |||
| BC009648.1 | - | 13 | 1437 | 3454326 | ||
| BC025315.1 | - | 13 | 1415 | 5458850 | ||
| BX537655.1 | - | 12 | 1394 | |||

WDR70
Genomic Map