siRNA Details
| Gene Symbol: RBM28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018077.2 | NP_060547.2 | 3 | 463 | ||
| XM_011516370.2 | XP_011514672.1 | 4 | 570 | |||
| XM_017012389.1 | XP_016867878.1 | 4 | 571 | |||
| XM_017012390.1 | XP_016867879.1 | 3 | 463 | |||
| GenBank mRNA | AK001239.1 | - | 3 | 463 | ||
| AK222716.1 | - | 3 | 425 | |||
| BC013889.2 | - | 3 | 417 | 3926178 | ||
| DA566130.1 | - | 3 | 463 | |||
| KU178685.1 | - | 3 | 348 | |||

RBM28
Genomic Map