siRNA Details
| Gene Symbol: UBA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018227.5 | NP_060697.4 | 5 | 352 | ||
| XM_017008359.1 | XP_016863848.1 | 5 | 329 | |||
| GenBank mRNA | AB014773.1 | - | 5 | 300 | ||
| AK094969.1 | - | 5 | 329 | |||
| AK314371.1 | - | 5 | 329 | |||
| AL596839.1 | - | 5 | 380 | |||
| AL832015.2 | - | 5 | 321 | |||
| AL832458.2 | - | 5 | 321 | |||
| BC031637.1 | - | 5 | 334 | |||
| BC126484.1 | - | 5 | 347 | 8992200 | ||
| BC126486.1 | - | 5 | 347 | 8992202 | ||
| EF623993.1 | - | 5 | 300 | |||

UBA6
Genomic Map