siRNA Details
| Gene Symbol: TMEM184C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018241.2 | NP_060711.2 | 6 | 1159 | ||
| GenBank mRNA | AK000169.1 | - | 3 | 312 | ||
| AK001708.1 | - | 6 | 1102 | |||
| BC046128.1 | - | 6 | 1161 | 5750613 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM184C| Gene Symbol: TMEM184C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018241.2 | NP_060711.2 | 6 | 1159 | ||
| GenBank mRNA | AK000169.1 | - | 3 | 312 | ||
| AK001708.1 | - | 6 | 1102 | |||
| BC046128.1 | - | 6 | 1161 | 5750613 | ||