siRNA Details
| Gene Symbol: C7orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303470.1 | NP_001290399.1 | 2, 3 | 226 | ||
| NM_018275.4 | NP_060745.3 | 2, 3 | 726 | |||
| GenBank mRNA | AK054872.1 | - | 2, 3 | 222 | ||
| AK310170.1 | - | 3, 4 | 490 | |||
| BC002749.2 | - | 2, 3 | 126 | |||
| BC015722.2 | - | 2, 3 | 684 | 3920412 | ||

C7orf43
Genomic Map